R&D Systems
Manufacturer Code:MAB10042100
Catalog # MAB10042100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 995234 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human CETP Cys18-Ser493 Accession # P11597 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: BPI fold containing family F; BPIFF cholesteryl ester transfer protein cholesteryl ester transfer protein plasma HDLCQ10 Lipid Transfer Protein I; Cholesteryl ester transfer protein; cholesteryl ester transfer protein plasma; cholesteryl ester transfer protein, plasma; Lipid transfer protein I
Gene Aliases: BPIFF; CETP; HDLCQ10
UniProt ID: (Human) P11597
Entrez Gene ID: (Human) 1071
Molecular Function: antibacterial response protein defense/immunity protein transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.