Abnova
Manufacturer Code:PAB15901
Catalog # 89-117-947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S688 of <i>Arabidopsis thaliana</i> CESA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CESA1.
For Research Use Only
Protein Aliases: AtCesA1; AtRSW1; CELLULOSE SYNTHASE 1; Cellulose synthase A catalytic subunit 1 [UDP-forming]; F8B4.110; F8B4_110; Protein RADIALLY SWOLLEN 1; RADIALLY SWOLLEN 1; RSW1
Gene Aliases: anisotropy1; ANY1; AT4G32410; AtCESA1; CELLULOSE SYNTHASE 1; CESA1; F8B4.110; F8B4_110; RADIALLY SWOLLEN 1; RSW1
UniProt ID: (Arabidopsis) O48946
Entrez Gene ID: (Arabidopsis) 829376
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.