Invitrogen
Manufacturer Code:PA587139
Catalog # PIPA587139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CES3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 3; carboxylesterase 3 (brain); carboxylesterase 3, pseudogene 1; Carboxylesterase 3A; ES-male; Es-male carboxylesterase; ES31; esterase 31; Esterase-31; Liver carboxylesterase 31; Liver carboxylesterase 31 homolog
Gene Aliases: CES3; Ces3-ps1; Ces31; Ces3a; ES31; UNQ869/PRO1887
UniProt ID: (Human) Q6UWW8, (Mouse) Q63880
Entrez Gene ID: (Human) 23491, (Mouse) 382053, (Rat) 689754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.