Invitrogen
Manufacturer Code:PA559388
Catalog # PIPA559388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CES3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 3; carboxylesterase 3 (brain); CES3 carboxylesterase 3 carboxylesterase 3 (brain) EC 3.1.1 EC 3.1.1.1 ES31FLJ21736 esterase 31 Liver carboxylesterase 31 homolog; esterase 31; Liver carboxylesterase 31 homolog
Gene Aliases: CES3; ES31; UNQ869/PRO1887
UniProt ID: (Human) Q6UWW8
Entrez Gene ID: (Human) 23491
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.