Invitrogen
Manufacturer Code:PA547802
Catalog # PIPA547802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Carboxylesterase3/CES3 Tyr19-Glu561 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxyesterase ES-10; Carboxylesterase 1D; Carboxylesterase 3; CES3 carboxylesterase 3 carboxylesterase 3 (brain) EC 3.1.1 EC 3.1.1.1 ES31FLJ21736 esterase 31 Liver carboxylesterase 31 homolog; ES-HVEL; FAEE synthase; Fatty acid ethyl ester synthase; Liver carboxylesterase 10; pI 6.1 esterase; TGH; Triacylglycerol hydrolase
Gene Aliases: Ces1; Ces1d; Ces3; TGH
UniProt ID: (Mouse) Q8VCT4, (Rat) P16303
Entrez Gene ID: (Mouse) 104158, (Rat) 113902
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.