Invitrogen
Manufacturer Code:PA5100228
Catalog # PIPA5100228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CES3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxyesterase ES-10 carboxylesterase 1D Carboxylesterase 3 carboxylesterase 3 (brain) carboxylesterase 3 pseudogene 1 carboxylesterase 3a Ces1 Ces1d CES3 Ces31 Ces3a Ces3-ps1 EC 3.1.1 EC 3.1.1.1 Es31 ES31FLJ21736 ES-HVEL ES-male Es-male carboxylesterase esterase 31 esterase-31 FAEE synthase fatty acid ethyl ester synthase Liver carboxylesterase 10 liver carboxylesterase 31 Liver carboxylesterase 31 homolog pI 6.1 esterase TGH Triacylglycerol hydrolase UNQ869/PRO1887; Carboxylesterase 1D; Carboxylesterase 3; carboxylesterase 3 (brain); Carboxylesterase 3A; ES-male; Es-male carboxylesterase; esterase 31; Esterase-31; FAEE synthase; Fatty acid ethyl ester synthase; Liver carboxylesterase 31; Liver carboxylesterase 31 homolog; TGH; Triacylglycerol hydrolase
Gene Aliases: Ces1; Ces1d; CES3; Ces31; Ces3a; ES31; TGH; UNQ869/PRO1887
UniProt ID: (Human) Q6UWW8, (Mouse) Q8VCT4, (Mouse) Q63880
Entrez Gene ID: (Human) 23491, (Mouse) 104158, (Mouse) 382053
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.