Proteintech
Manufacturer Code:145871AP150UL
Catalog # 50-172-9563
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CES3 Fusion Protein Ag6113 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 1D; Carboxylesterase 3; carboxylesterase 3 (brain); carboxylesterase 3 CES3 ES31; esterase 31; FAEE synthase; Fatty acid ethyl ester synthase; Liver carboxylesterase 31 homolog; TGH; Triacylglycerol hydrolase
Gene Aliases: Ces1; Ces1d; CES3; ES31; TGH; UNQ869/PRO1887
UniProt ID: (Human) Q6UWW8, (Mouse) Q8VCT4
Entrez Gene ID: (Human) 23491, (Mouse) 104158
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.