Invitrogen
Manufacturer Code:PA581353
Catalog # PIPA581353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse CES2/Carboxylesterase 2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; Carboxylesterase 2; carboxylesterase 2A; carboxylesterase 2E; carboxylesterase 5; carboxylesterase 6; carboxylesterase ML3; Carboxylic ester hydrolase; Carboxylic ester hydrolase 2A; CE-2 CES2A1 PCE-2 iCE carboxylesterase 2 (intestine liver) cocaine esterase hCE-2 intestinal carboxylesterase liver carboxylesterase-2 methylumbelliferyl-acetate deacetylase 2; CES 2; Peroxisome proliferator-inducible acylcarnitine hydrolase; Pyrethroid hydrolase Ces2a; Pyrethroid hydrolase Ces2e
Gene Aliases: 9030624L02Rik; 9130231C15Rik; ACH M1; AI266984; CES 2; Ces2; Ces2a; ces2A3; Ces2c; Ces2e; Ces5; Ces6
UniProt ID: (Mouse) Q8QZR3, (Mouse) Q91WG0, (Mouse) Q8BK48
Entrez Gene ID: (Mouse) 102022, (Mouse) 234671, (Mouse) 234673
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.