Invitrogen
Manufacturer Code:PA576083
Catalog # PIPA576083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CES2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 2; carboxylesterase 2 (intestine, liver); carboxylesterase 2A; carboxylesterase 6; Carboxylic ester hydrolase 2A; CE-2; Cocaine esterase; Cocaine esterase Carboxylesterase 2 CE-2 hCE-2 Methylumbelliferyl-acetate deacetylase 2 CES-2 ICE; hCE-2; intestinal carboxylesterase; liver carboxylesterase-2; Methylumbelliferyl-acetate deacetylase 2; Pyrethroid hydrolase Ces2a
Gene Aliases: 9130231C15Rik; AI266984; CE-2; CES2; Ces2a; CES2A1; Ces6; ICE; PCE-2
UniProt ID: (Human) O00748, (Mouse) Q8QZR3
Entrez Gene ID: (Human) 8824, (Mouse) 102022
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.