Invitrogen
Manufacturer Code:PA547715
Catalog # PIPA547715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Carboxylesterase 2/CES2 Gln27-Lys557 (predicted) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; Carboxylesterase 2; carboxylesterase ML3; Carboxylic ester hydrolase; CE-2 CES2 PCE-2; CES 2; Peroxisome proliferator-inducible acylcarnitine hydrolase
Gene Aliases: ACH M1; CES 2; Ces2; ces2A3; Ces2c
UniProt ID: (Mouse) Q91WG0
Entrez Gene ID: (Mouse) 234671
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.