Invitrogen
Manufacturer Code:PA5102415
Catalog # PIPA5102415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CES2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9030624L02Rik ACH M1 Acylcarnitine hydrolase acylcarnitine hydrolase M1 carboxylesterase 2 carboxylesterase 2 (intestine liver) carboxylesterase 2C carboxylesterase 2E carboxylesterase 5 carboxylesterase ML3 CE-2 CES 2 Ces2 CES-2 CES2A1 ces2A3 Ces2c Ces2e Ces2l Ces5 CESRL4 cocaine esterase hCE-2 ICE intestinal carboxylesterase liver carboxylesterase-2 methylumbelliferyl-acetate deacetylase 2 P iCE PCE-2 Peroxisome proliferator-inducible acylcarnitine hydrolase phenobarbital-inducible carboxylesterase (liver) pyrethroid hydrolase Ces2e; ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; Carboxylesterase 2; carboxylesterase 2 (intestine, liver); carboxylesterase 2C; carboxylesterase 2E; carboxylesterase 5; carboxylesterase ML3; Carboxylic ester hydrolase; CE-2; CES 2; Cocaine esterase; hCE-2; intestinal carboxylesterase; liver carboxylesterase-2; Methylumbelliferyl-acetate deacetylase 2; Peroxisome proliferator-inducible acylcarnitine hydrolase; phenobarbital-inducible carboxylesterase (liver); Pyrethroid hydrolase Ces2e
Gene Aliases: 9030624L02Rik; ACH M1; CE-2; CES 2; CES2; CES2A1; ces2A3; Ces2c; Ces2e; Ces2l; Ces5; CESRL4; ICE; PCE-2
UniProt ID: (Human) O00748, (Rat) O70631, (Mouse) Q91WG0, (Mouse) Q8BK48
Entrez Gene ID: (Human) 8824, (Rat) 100910144, (Rat) 171118, (Mouse) 234671, (Mouse) 234673, (Rat) 192257
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.