Proteintech
Manufacturer Code:153781AP150UL
Catalog # 50-173-0188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CES2 Fusion Protein Ag2218 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carboxylesterase 2; carboxylesterase 2 (intestine, liver); Carboxylesterase 2 CE 2 CES2 CES2A1 hCE 2 iCE PCE 2; CE-2; Cocaine esterase; hCE-2; intestinal carboxylesterase; liver carboxylesterase-2; Methylumbelliferyl-acetate deacetylase 2
Gene Aliases: CE-2; CES2; CES2A1; ICE; PCE-2
UniProt ID: (Human) O00748
Entrez Gene ID: (Human) 8824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.