Invitrogen
Manufacturer Code:PA579032
Catalog # PIPA579032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human CES1 recombit protein (Position: E99-A206). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; ES-HTEL; ES-x; esterase 22; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Liver carboxylesterase 1 Acyl-coenzyme A:cholesterol acyltransferase ACAT Brain carboxylesterase hBr1 Carboxylesterase 1 CE-1 hCE-1; liver carboxylesterase 3; Methylumbelliferyl-acetate deacetylase 1; Monocyte/macrophage serine esterase; pI 5.5 esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; Ces-1; CES1; Ces1g; CES2; Es22; hCE-1; HMSE; HMSE1; PCE-1; REH; Ses-1; SES1; TGH
UniProt ID: (Human) P23141, (Mouse) Q8VCC2
Entrez Gene ID: (Human) 1066, (Rat) 29225, (Mouse) 12623
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.