Invitrogen
Manufacturer Code:PA519740
Catalog # PIPA519740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 500 to the C-terminus of Human Liver Carboxylesterase 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT; ACAT CEH CES2 HMSE HMSE1 MGC117365 PCE-1 SES1 TGH REH acyl coenzyme A:cholesterol acyltransferase acyl-coenzyme A:cholesterol acyltransferase brain carboxylesterase hBr1 carboxylesterase 1 (monocyte/macrophage serine esterase 1) carboxylesterase 2 (liver) cholesteryl ester hydrolase cocaine carboxylesterase egasyn human monocyte/macrophage serine esterase 1 liver carboxylesterase 1 monocyte/macrophage serine esterase retinyl ester hydrolase serine esterase 1 triacylglycero; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1-like; Carboxylesterase 1G; carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; ES-x; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Methylumbelliferyl-acetate deacetylase 1; Monocyte/macrophage serine esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; Ces-1; CES1; Ces1g; Ces1l; CES2; EG244595; Gm4976; hCE-1; HMSE; HMSE1; PCE-1; REH; Ses-1; SES1; TGH
UniProt ID: (Human) P23141, (Mouse) Q8VCC2
Entrez Gene ID: (Human) 1066, (Rat) 679817, (Mouse) 244595, (Mouse) 12623
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.