Invitrogen
Manufacturer Code:PA5112874
Catalog # PIPA5112874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Mouse Carboxylesterase 1C protein (19-550AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACAT; ACAT acyl coenzyme A:cholesterol acyltransferase Acyl-coenzyme A:cholesterol acyltransferase Brain carboxylesterase hBr1 carboxyesterase ES-3 Carboxylesterase 1 carboxylesterase 1 (monocyte/macrophage serine esterase 1) carboxylesterase 1A Carboxylesterase 1E Carboxylesterase 1G carboxylesterase 1-like carboxylesterase 2 (liver) CE-1 CEH CES1 Ces-1 Ces1a Ces1e Ces1g Ces1l CES2 cholesteryl ester hydrolase Cocaine carboxylesterase Eg EG244595 Egasyn Es22 Es-22 ES-HTEL esterase 22 esterase-22 ES-x Gm4976 hCE-1 HMSE HMSE1 human monocyte/macrophage serine esterase 1 Liver carboxylesterase 1 Liver carboxylesterase 22 liver carboxylesterase 3 Methylumbelliferyl-acetate deacetylase 1 MGC117365 Monocyte/macrophage serine esterase PCE-1 pI 5.5 esterase REH Retinyl ester hydrolase Serine esterase 1 SES1 Ses-1 TGH triacylglycero Triacylglycerol hydrolase; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1-like; Carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; Es-22; ES-HTEL; ES-x; esterase 22; esterase-22; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; Methylumbelliferyl-acetate deacetylase 1; Monocyte/macrophage serine esterase; pI 5.5 esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; Ces-1; CES1; Ces1e; Ces1g; Ces1l; CES2; Eg; EG244595; egasyn; Es-22; Es22; Gm4976; hCE-1; HMSE; HMSE1; PCE-1; REH; Ses-1; SES1; TGH
UniProt ID: (Human) P23141, (Mouse) Q64176, (Mouse) Q8VCC2
Entrez Gene ID: (Human) 1066, (Rat) 679817, (Mouse) 244595, (Rat) 29225, (Mouse) 13897, (Mouse) 12623
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.