Invitrogen
Manufacturer Code:PA5115035
Catalog # PIPA5115035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human COL4A3BP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810404O15Rik 9230101K08Rik alpha 3 type IV collagen binding protein AU016711 ceramide transfer protein ceramide transport protein ceramide transporter CERT CERTL Col4a3bp collagen type IV alpha 3 binding protein collagen type IV alpha-3-binding protein collagen type IV alpha 3 (Goodpasture antigen) binding protein goodpasture antigen-binding protein GPBP hCERT lipid-transfer protein CERTL MRD34 procollagen type IV alpha 3 (Goodpasture antigen) binding protein STARD11 StAR-related lipid transfer (START) domain containing 11 stAR-related lipid transfer protein 11 START domain-containing protein 11; alpha 3 type IV collagen binding protein; Ceramide transfer protein; ceramide transport protein; ceramide transporter; CERT; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: 2810404O15Rik; 9230101K08Rik; AU016711; CERT; CERT1; CERTL; COL4A3BP; GPBP; MRD34; STARD11
UniProt ID: (Human) Q9Y5P4, (Mouse) Q9EQG9
Entrez Gene ID: (Human) 10087, (Mouse) 68018, (Rat) 365652
Molecular Function: kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.