Invitrogen
Manufacturer Code:PA532707
Catalog # PIPA532707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human CERK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; hCERK; Lipid kinase 4; lipid kinase LK4; LK4; LK4 dA59H18.2 dA59H18.3 hCERK acylsphingosine kinase lipid kinase 4 lipid kinase LK4
Gene Aliases: CERK; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4
UniProt ID: (Human) Q8TCT0
Entrez Gene ID: (Human) 64781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.