Invitrogen
Manufacturer Code:PA513856
Catalog # PIPA513856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 487-516 amino acids from the C-terminal region of human CERK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; Ceramide kinase Acylsphingosine kinase hCERK Lipid kinase 4 LK4 KIAA1646; hCERK; Lipid kinase 4; lipid kinase LK4; LK4; mCERK
Gene Aliases: AI848610; CERK; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4; mKIAA1646
UniProt ID: (Human) Q8TCT0, (Mouse) Q8K4Q7
Entrez Gene ID: (Human) 64781, (Mouse) 223753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.