Abnova
Manufacturer Code:PAB4648
Catalog # 89-131-512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CERK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CERK.
For Research Use Only
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; dA59H18.2; dA59H18.3; DKFZp434E0211; FLJ21430; FLJ23239; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; MGC131878
Gene Aliases: CERK; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4
UniProt ID: (Human) Q8TCT0
Entrez Gene ID: (Human) 64781
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.