Invitrogen
Manufacturer Code:MA515554
Catalog # PIMA515554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9D6 |
Immunogen | Purified recombit fragment of human CER1 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cerberus; cerberus 1, cysteine knot superfamily, homolog; cerberus-related 1; Cerberus-related protein; DAN domain family member 4; DAND4 MGC96951 MGC119894 MGC119895 CER1
Gene Aliases: CER1; DAND4
UniProt ID: (Human) O95813
Entrez Gene ID: (Human) 9350
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.