Invitrogen
Manufacturer Code:PA523876
Catalog # PIPA523876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 29-57 amino acids from the N-terminal region of human CEPT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: choline/ethanolaminephosphotransferase; Choline/ethanolaminephosphotransferase 1; choline/ethanolaminephosphotransferase choline/ethanolaminephosphotransferase 1 hCEPT1; hCEPT1
Gene Aliases: CEPT1; PRO1101
UniProt ID: (Human) Q9Y6K0
Entrez Gene ID: (Human) 10390
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.