Proteintech
Manufacturer Code:204961AP150UL
Catalog # 50-173-2921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CEPT1 Fusion Protein Ag14140 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEPT1 hCEPT1; choline/ethanolaminephosphotransferase; Choline/ethanolaminephosphotransferase 1; hCEPT1; mCEPT1
Gene Aliases: 9930118K05Rik; CEPT1; mCEPT1; PRO1101
UniProt ID: (Human) Q9Y6K0, (Rat) Q6AXM5, (Mouse) Q8BGS7
Entrez Gene ID: (Human) 10390, (Rat) 310773, (Mouse) 99712
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.