Proteintech
Manufacturer Code:220501AP150UL
Catalog # 50-173-3704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CEP97 Fusion Protein Ag16995 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810403B08Rik centrosomal protein 97kDa Centrosomal protein of 97 kDa CEP97 LRRIQ2; centrosomal protein 97kDa; Centrosomal protein of 97 kDa; Cep97; Leucine-rich repeat and IQ domain-containing protein 2; leucine-rich repeats and IQ motif containing 2
Gene Aliases: 2810403B08Rik; CEP97; LRRIQ2
UniProt ID: (Human) Q8IW35
Entrez Gene ID: (Dog) 608540, (Human) 79598
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.