Proteintech
Manufacturer Code:144981AP150UL
Catalog # 50-172-9493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CEP250/CNAP1 Fusion Protein Ag5925 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 250 kDa centrosomal protein; 250 kDa centrosomal protein C NAP1 Centrosomal protein 2 centrosomal protein 250kDa CEP2 CEP250 CNAP1 C-NAP1; C-Nap1; Centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250kDa; centrosome associated protein; Centrosome-associated protein CEP250; Cep250
Gene Aliases: C-NAP1; CEP2; CEP250; CNAP1
UniProt ID: (Human) Q9BV73
Entrez Gene ID: (Human) 11190
Molecular Function:
enzyme modulator
kinase modulator
viral protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.