Proteintech
Manufacturer Code:218151AP150UL
Catalog # 50-173-3598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: asterless; centrosomal protein 152kDa; centrosomal protein 152kDa Centrosomal protein of 152 kDa CEP152 KIAA0912; Centrosomal protein of 152 kDa; Cep152; microcephaly, primary autosomal recessive 4
Gene Aliases: AI851464; CEP152; KIAA0912; MCPH4; MCPH9; mKIAA0912; SCKL5
UniProt ID: (Human) O94986, (Mouse) A2AUM9
Entrez Gene ID: (Human) 22995, (Mouse) 99100
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.