Invitrogen
Manufacturer Code:PA536440
Catalog # PIPA536440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1087-1140 of human CEP135 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: centrosomal protein 135 kDa centrosomal protein 135kDa Centrosomal protein 4 Centrosomal protein of 135 kDa centrosome protein 4 centrosome protein cep135 Cep135 Cep135 CEP4; centrosomal protein 135kDa; Centrosomal protein 4; Centrosomal protein of 135 kDa; centrosome protein cep135; Cep135
Gene Aliases: BC062951; CEP135; CEP4; Gm1044; KIAA0635; MCPH8
UniProt ID: (Human) Q66GS9, (Mouse) Q6P5D4
Entrez Gene ID: (Human) 9662, (Mouse) 381644
Molecular Function:
enzyme modulator
kinase modulator
viral protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.