Invitrogen
Manufacturer Code:PA5101507
Catalog # PIPA5101507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CEP135 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC062951 centrosomal protein 135 centrosomal protein 135kDa Centrosomal protein 4 centrosomal protein of 135 kDa centrosome protein cep135 CEP135 CEP4 Gm1044 KIAA0635 MCPH8 RGD1310726; centrosomal protein 135kDa; Centrosomal protein 4; Centrosomal protein of 135 kDa; centrosome protein cep135; Cep135
Gene Aliases: BC062951; CEP135; CEP4; Gm1044; KIAA0635; MCPH8
UniProt ID: (Human) Q66GS9, (Mouse) Q6P5D4
Entrez Gene ID: (Human) 9662, (Mouse) 381644
Molecular Function:
enzyme modulator
kinase modulator
viral protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.