Invitrogen
Manufacturer Code:PA524429
Catalog # PIPA524429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 850-877 amino acids from the C-terminal region of human CENTG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGAP-2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin, gamma 1; Centaurin-gamma-1; CENTG1 GGAP2 PIKE Arf GAP with GTP-binding protein-like ANK repeat and PH domains 2 GTP-binding and GTPase activating protein 2 arf-GAP with GTPase ANK repeat and PH domain-containing protein 2 centaurin gamma 1 cnt-g1 phosphatidylinositol-3-kinase enhancer phosphoinosi; Cnt-g1; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; Phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; PIKE
Gene Aliases: AGAP-2; AGAP2; CENTG1; cnt-g1; GGAP2; KIAA0167; mKIAA0167; PIKE
UniProt ID: (Human) Q99490, (Mouse) Q3UHD9
Entrez Gene ID: (Human) 116986, (Mouse) 216439
Molecular Function: G-protein modulator enzyme modulator nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.