Novus Biologicals
Manufacturer Code:NBP292377100UL
Catalog # NB026635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 180-300 of human CENPN (NP_001094094). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM039 C16orf60 CENP-N centromere protein N chromosome 16 open reading frame 60 FLJ13607 FLJ22660 ICEN32 Interphase centromere complex protein 32; CENP-N; Centromere protein N; Interphase centromere complex protein 32
Gene Aliases: BM-309; BM039; C16orf60; CENP-N; CENPN; ICEN32
UniProt ID: (Human) Q96H22
Entrez Gene ID: (Human) 55839
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.