Proteintech
Manufacturer Code:167511AP150UL
Catalog # 50-173-1157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CENPN Fusion Protein Ag10219 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM039 C16orf60 CENP N CENPN centromere protein N ICEN32; CENP-N; Centromere protein N; Interphase centromere complex protein 32
Gene Aliases: 2610510J17Rik; AI426416; AW545024; BM-309; BM039; C16orf60; CENP-N; CENPN; Da2-27; Da2-4; ICEN32; RGD1310953
UniProt ID: (Human) Q96H22, (Rat) Q5U2W4, (Mouse) Q9CZW2
Entrez Gene ID: (Human) 55839, (Rat) 361416, (Mouse) 72155
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.