Proteintech
Manufacturer Code:115171AP150UL
Catalog # 50-556-958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CENPJ Fusion Protein Ag2078 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM032 CENP J CENPJ centromere protein J CPAP LAG 3 associated protein LAP LIP1 LYST interacting protein 1 MCPH6; CENP-J; Centromere protein J; Centrosomal P4.1-associated protein; LAG-3-associated protein; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7
Gene Aliases: 4932437H03Rik; BM032; CENP-J; CENPJ; CPAP; Gm81; LAP; LIP1; MCPH6; Sas-4; Sas4; SASS4; SCKL4
UniProt ID: (Human) Q9HC77, (Mouse) Q569L8
Entrez Gene ID: (Human) 55835, (Rat) 305909, (Mouse) 219103
Molecular Function: receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.