Novus Biologicals
Manufacturer Code:NB500101SS
Catalog # NB500101SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A bacterial fusion protein from the C-terminus of human CENPF. [UniProt# P49454] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AH antigen; AH antigen cell-cycle-dependent 350K nuclear protein CENF CENP-F CENP-F kinetochore protein centromere protein F centromere protein F (350/400kD mitosin) centromere protein F 350/400ka (mitosin) centromere protein F 350/400kDa (mitosin) hcp-1 Kinetochore protein CENPF mitosin PRO1779; cell-cycle-dependent 350K nuclear protein; CENP-F; CENP-F kinetochore protein; Centromere protein F; centromere protein F, 350/400ka (mitosin); centromere protein F, 350/400kDa (mitosin); Kinetochore protein CENPF; Mitosin
Gene Aliases: CENF; CENPF; CILD31; hcp-1; PRO1779; STROMS
UniProt ID: (Human) P49454
Entrez Gene ID: (Human) 1063
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.