Proteintech
Manufacturer Code:281421AP150UL
Catalog # 50-173-5820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CENPE Fusion Protein Ag27230 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CENP E CENPE Centromere protein E centromere protein E 312kDa KIF10 Kinesin related protein CENPE; CENP-E; Centromere autoantigen E (312kD); Centromere protein E; centromere protein E, 312kDa; Centromere-associated protein E; kinesin family member 10; Kinesin-7; Kinesin-related protein CENPE; protein phosphatase 1, regulatory subunit 61
Gene Aliases: CENP-E; CENPE; KIF10; MCPH13; PPP1R61
UniProt ID: (Human) Q02224
Entrez Gene ID: (Human) 1062
Molecular Function: cytoskeletal protein microtubule binding motor protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.