Novus Biologicals
Manufacturer Code:NBP170495
Catalog # NBP170495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CEND1(cell cycle exit and neuronal differentiation 1) The peptide sequence was selected from the N terminal of CEND1. Peptide sequence MESRGKSASSPKPDTKVPQVTTEAKVPPAADGKAPLTKPSKKEAPAEKQQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM88 antigen; BM88 antigen cell cycle exit and neuronal differentiation 1 cell cycle exit and neuronal differentiation protein 1 FLJ90066; Cell cycle exit and neuronal differentiation protein 1
Gene Aliases: BM88; CEND1
UniProt ID: (Human) Q8N111
Entrez Gene ID: (Human) 51286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.