Invitrogen
Manufacturer Code:PA568396
Catalog # PIPA568396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human CEND. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM88 antigen; BM88 BM88 antigen cell cycle exit and neuronal differentiation protein 1; Cell cycle exit and neuronal differentiation protein 1
Gene Aliases: 1500001H12Rik; AI415214; BM88; C38; CEND1; RGD1309401
UniProt ID: (Human) Q8N111, (Rat) Q5FVI4, (Mouse) Q9JKC6
Entrez Gene ID: (Human) 51286, (Rat) 361675, (Mouse) 57754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.