Invitrogen
Manufacturer Code:PA538870
Catalog # PIPA538870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the N-terminal region of human CEND |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BM88 antigen; BM88 BM88 antigen cell cycle exit and neuronal differentiation protein 1; Cell cycle exit and neuronal differentiation protein 1
Gene Aliases: BM88; CEND1
UniProt ID: (Human) Q8N111
Entrez Gene ID: (Human) 51286
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.