Abnova
Manufacturer Code:PAB26118
Catalog # 89-127-759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at C-terminus of human CELSR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CELSR3.
For Research Use Only
Protein Aliases: adhesion G protein-coupled receptor C3; anchor protein; Cadherin EGF LAG seven-pass G-type receptor 3; Cadherin family member 11; cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila); CDHF11; EGF-like protein 1; EGF-like-domain, multiple 1; EGFL1; Epidermal growth factor-like protein 1; Flamingo homolog 1; FMI1; HFMI1; MEGF2; Multiple EGF-like domains protein 2; Multiple epidermal growth factor-like domains protein 2; RESDA1
Gene Aliases: ADGRC3; CDHF11; CELSR3; EGFL1; FMI1; HFMI1; KIAA0812; MEGF2; RESDA1
UniProt ID: (Human) Q9NYQ7
Entrez Gene ID: (Human) 1951
Molecular Function: G-protein coupled receptor cadherin cell adhesion molecule receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.