Novus Biologicals
Manufacturer Code:NBP256650
Catalog # NBP256650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KHLKGVLGGRKLHLEDSATTRATLLTRSLNCNTTFGDGPDMLRTDLGESTASLDSIVRDEGIQKLGVSSGLVRGSHGEPDASLMPRSCKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adhesion G protein-coupled receptor C1; Cadherin EGF LAG seven-pass G-type receptor 1; cadherin EGF LAG seven-pass G-type receptor 1 (flamingo homolog Drosophila) DKFZp434P0729 flamingo homolog 2 FMI2 HFMI2 ME2 protocadherin flamingo 2; Cadherin family member 9; cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila); Flamingo homolog 2; hFmi2; protocadherin flamingo 2
Gene Aliases: ADGRC1; CDHF9; CELSR1; FMI2; HFMI2; ME2
UniProt ID: (Human) Q9NYQ6
Entrez Gene ID: (Human) 9620
Molecular Function:
G-protein coupled receptor
cadherin
cell adhesion molecule
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.