Abnova
Manufacturer Code:PAB26153
Catalog # 89-127-793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at N-terminus of human CELSR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CELSR1.
For Research Use Only
Protein Aliases: adhesion G protein-coupled receptor C1; Cadherin EGF LAG seven-pass G-type receptor 1; Cadherin family member 9; cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila); CDHF9; DKFZp434P0729; Flamingo homolog 2; FMI2; HFMI2; ME2; protocadherin flamingo 2
Gene Aliases: ADGRC1; CDHF9; CELSR1; FMI2; HFMI2; ME2
UniProt ID: (Human) Q9NYQ6
Entrez Gene ID: (Human) 9620
Molecular Function:
G-protein coupled receptor
cadherin
cell adhesion molecule
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.