Invitrogen
Manufacturer Code:PA552504
Catalog # PIPA552504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CEL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bile salt-dependent lipase, oncofetal isoform; bucelipase; carboxyl ester hydrolase; carboxyl ester lipase (bile salt-stimulated lipase); Carboxyl ester lipase (bile salt-stimulated lipase) BAL BSDL BSSL CELL CEase FAP FAPP LIPA MODY8; cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; sterol esterase
Gene Aliases: BAL; BSDL; BSSL; CEase; CELL; FAP; FAPP; LIPA; MODY8
Entrez Gene ID: (Human) 1056
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.