Invitrogen
Manufacturer Code:PA540793
Catalog # PIPA540793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CEL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; Bile salt-stimulated lipase; BSSL; bucelipase; carboxyl ester hydrolase; Carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); Carboxyl ester lipase (bile salt-stimulated lipase) BAL BSDL BSSL CELL CEase FAP FAPP LIPA MODY8; Cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; Pancreatic lysophospholipase; Sterol esterase
Gene Aliases: 1810036E18Rik; BAL; BSDL; BSSL; CEase; Cel; CELL; FAP; FAPP; Lip1; LIPA; MODY8
UniProt ID: (Mouse) Q64285
Entrez Gene ID: (Horse) 100066592, (Human) 1056, (Pig) 100625953, (Mouse) 12613, (Rat) 24254
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.