Invitrogen
Manufacturer Code:PA5115005
Catalog # PIPA5115005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CEL corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810036E18Rik BAL Bile salt-activated lipase bile salt-dependent lipase oncofetal isoform bile salt-stimulated lipase BSDL BSSL bucelipase carboxyl ester hydrolase carboxyl ester lipase carboxyl ester lipase (bile salt-stimulated lipase) CEase Cel CELL Cholesterol esterase FAP FAPP fetoacinar pancreatic protein Lip1 LIPA lysophospholipase pancreatic MODY8 Pancreatic lysophospholipase Sterol esterase; BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; Bile salt-stimulated lipase; BSSL; bucelipase; carboxyl ester hydrolase; Carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); Cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; Pancreatic lysophospholipase; Sterol esterase
Gene Aliases: 1810036E18Rik; BAL; BSDL; BSSL; CEase; Cel; CELL; FAP; FAPP; Lip1; LIPA; MODY8
UniProt ID: (Mouse) Q64285
Entrez Gene ID: (Human) 1056, (Mouse) 12613
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.