Novus Biologicals
Manufacturer Code:42100002
Catalog # 42-100-002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | In vivo generated recominant protein fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ced-10 Protein CED-10; Cell death protein 10; Cell-corpse engulfment protein ced-10; CErac1; GTPase; Ras-related protein ced-10; Ras-related protein rac-1
Gene Aliases: C09G12.8; ced-10; CELE_C09G12.8; rac-1
UniProt ID: (C. elegans) Q03206
Entrez Gene ID: (C. elegans) 177111
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.