Proteintech
Manufacturer Code:666491IG150UL
Catalog # 50-173-7067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B6E10 |
Immunogen | CEBPB Fusion Protein Ag20073 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C/EBP beta; C/EBP beta CEBPB CRP2 IL6DBP LAP Liver activator protein NF IL6 Nuclear factor NF IL6 TCF 5 TCF5 Transcription factor 5; C/EBP-related protein 2; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer-binding protein beta; IL-6DBP; interleukin 6-dependent DNA-binding protein; Interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; SF-B; Silencer factor B; TCF-5; Transcription factor 5; transcription factor C/EBP beta
Gene Aliases: C/EBP-beta; CEBPB; Crp2; IL6DBP; NF-IL6; PP9092; Sfb; TCF5
UniProt ID: (Human) P17676, (Rat) P21272
Entrez Gene ID: (Human) 1051, (Rat) 24253
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.