Novus Biologicals
Manufacturer Code:NBP237497
Catalog # NBP237497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B7 |
Immunogen | Synthesized peptide of human CEBPA (AA: C-RKSRDKAKRNVETKV). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C/EBP alpha; C/EBP alpha C/EBP-alpha CCAAT/enhancer binding protein (C/EBP) alpha CCAAT/enhancer-binding protein alpha CEBP; CCAAT/enhancer binding protein (C/EBP), alpha; CCAAT/enhancer-binding protein alpha
Gene Aliases: C/EBP-alpha; CEBP; CEBPA
UniProt ID: (Human) P49715
Entrez Gene ID: (Human) 1050
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.