R&D Systems
Manufacturer Code:MAB22441100
Catalog # MAB22441100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 283324 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human CEACAM-1 Gln35-Gly428 Accession # P13688 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen CD66; antigen CD66 BGP1 BGP-1 BGP1BGPBiliary glycoprotein 1 BGPI Biliary Glycoprotein 1 biliary glycoprotein adhesion molecule carcinoembryonic antigen-related cell adhesion molecule 1 carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein) CD66a CD66a antigen Cea-1 CEACAM1 Hv-1 Hv-2 MHVR; BGP-1; Biliary glycoprotein 1; Carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); CD66a; CD66a antigen
Gene Aliases: BGP; BGP1; BGPI; CEACAM1
UniProt ID: (Human) P13688
Entrez Gene ID: (Human) 634
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily kinase membrane-bound signaling molecule protein kinase receptor signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.