Invitrogen
Manufacturer Code:PA514250
Catalog # PIPA514250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 50-77 amino acids from the N-terminal region of human CEACAM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen CD66; BGP BGP1 BGPI CD66a antigen antigen CD66 carcinoembryonic antigen-related cell adhesion molecule 1; BGP-1; Biliary glycoprotein 1; Carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); CD66a; CD66a antigen
Gene Aliases: BGP; BGP1; BGPI; CEACAM1
UniProt ID: (Human) P13688
Entrez Gene ID: (Human) 634
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
kinase
membrane-bound signaling molecule
protein kinase
receptor
signaling molecule
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.