OriGene
Manufacturer Code:TA803413
Catalog # 50-167-9734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 35-680 of human CEACAM5 produced in SF9 cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Carcinoembryonic antigen; Carcinoembryonic antigen-related cell adhesion molecule 5; CD66e; CD66e CEA; CEA; Meconium antigen 100
Gene Aliases: CD66e; CEA; CEACAM5
UniProt ID: (Human) P06731
Entrez Gene ID: (Human) 1048
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.