Invitrogen
Manufacturer Code:MA530116
Catalog # PIMA530116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6 |
Immunogen | Recombit Human ALK-6/BMPR1B protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activin receptor-like kinase 6; BMP type-1B receptor; BMPR-1B; bone morphogenetic protein receptor type IB; Bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type IB; CDw293; Serine(threonine) protein kinase activin receptor-like kinase 6 ALK6; serine/threonine receptor kinase
Gene Aliases: ALK-6; ALK6; AMDD; BDA1D; BDA2; BMPR1B; CDw293
UniProt ID: (Human) O00238
Entrez Gene ID: (Human) 658
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.