Invitrogen
Manufacturer Code:MA514494
Catalog # PIMA514494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | EPR2764Y |
Immunogen | A synthetic peptide corresponding to residues near the N-terminus of human CDX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeo box transcription factor 2 caudal type homeobox transcription factor 2 caudal-type homeobox protein 2 homeobox protein CDX-2; caudal type homeobox transcription factor 2; Caudal-type homeobox protein 2; CDX-3; Homeobox protein CDX-2; homeobox protein miniCDX2
Gene Aliases: Cdx-2; CDX-3; CDX2; CDX2/AS; CDX3
UniProt ID: (Human) Q99626, (Mouse) P43241
Entrez Gene ID: (Human) 1045, (Mouse) 12591
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.